ContactUsLinkComponent
HCV-NS3-S1b
HCV-NS3-S1b
Supplier:  Biorbyt
Certificates
HCV-NS3-S1b
Supplier:  Biorbyt
Specifications
  • Condizioni di conservazione:
    Store at−20 °C.
  • Protein/peptide name:
    HCV-NS3-S1b
  • Purezza:
    Greater than 95% by SDS-PAGE
  • Formulazione:
    Liquid. Supplied in 25 mM Tris-HCl pH 8.0, 5mMglutathione, 1.5 M urea, 0.2% Triton X-100 and 50%glycerol.
  • Applicazioni testate:
    ELISA , WB
Specifications
Related Information
Frequently Bought Together