Protein-A/G (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Sorgente luminosa:E. coli
- Condizioni di conservazione:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein/peptide name:Protein-A/G
- Purezza:>95% pure as determined by SDS-PAGE and RP-HPLC. A280 of 0.1% pure solution~0.505.
- Formulazione:Lyophilized white Powder containing no additives.
- Applicazioni testate:SDS-PAGE