Shoe rack
Shoe rack, wire frame Ø 8 mm, continuous wire Ø 5 mm, 263×180×600 mm
Codice catalogo HUPF7500452
Nome del fornitore: HUPFER METALLWERKE
Vai a
Product Details & Documents
This shoe rack is made of stainless steel with electropolished finish. It can hold 6 pairs of shoes or clogs suspended and also it can be fitted to wall-mounted support and trolleys.
Specifiche
- Descrizione:Shoe rack, wire frame Ø 8 mm, continuous wire Ø 5 mm
- L×P×A:263×180×600 mm