Orders LinkContactUsLinkComponent
Adhesive mounting tapes
Adhesive mounting tapes
Supplier:   ISOPAD
Adhesive mounting tapes
Supplier:  ISOPAD
Certificates
Adhesive mounting tapes
2 Options Available Below
Product Details & Documents
Glass-silk adhesive tapes used for mounting.
Product Family Options
Length 33 m
Product Information
  • Descrizione
    Larghezza
    Availability
    Price
  • Glass silk adhesive tape, max. 200 °C
    15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45.01 SFr.
    45.01 SFr.
    1 * 1 ST
       45.01 SFr.
Length 100 m
Product Information
  • Descrizione
    Larghezza
    Availability
    Price
  • Glass silk tape, max. 450 °C
    12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34.32 SFr.
    34.32 SFr.
    1 * 1 ST
       34.32 SFr.