Acidic cleaner and neutraliser, ProCare Lab 30 C
MIEL10266680
: MIELE
- Capacità:10 l
- Descrizione:Acidic cleaner, ProCare lab 30 C
- Seq:02
- Conf:10 L
- Valore pH:2,5 - 2,9
Liquid, acidic citric acid based cleaning concentrate for automated cleaning of laboratory glassware and reusable laboratory material.
- Good rinsing properties
- Free from tensides and phosphates
- Removes residues soluble in acids
ProCare Lab 30 C is intended for general use in medical, biological, microbiological and chemical laboratories, food-processing, cosmetics and pharmaceutical industries.
Note: Suitable for stainless steel, laboratory glassware, ceramics, plastics such as PE, PP, PVDF, PTFE and also for neutralisation of non anodised aluminium. Not suitable for use on non ferrous metals, light alloys, anodised aluminium, aluminium, chrome- and nickel-plated parts.