Red long-term cell tracer, CYTO-ID®
Codice catalogo ENZOENZ51037K025
Nome del fornitore: Enzo Life Sciences
Numero CAS:
Specifiche
- Conf:1 KIT
- Descrizione:Red long-term cell tracer
- Seq:02
Specifiche
Informazioni su questo articolo
CYTO-ID® is a live cell fluorescent labelling used for a long term cell viability, cytotoxicity, cell adhesion, cell migration, and cell-cell fusion studies.
- Dual labeling with CELLESTIAL® fluorescent probes
- Minimal transfer of fluorescence from dye-labeled to unlabelled cells
Uses proprietary non covalent cell labeling technology to stably incorporate a red fluorescent dye containing hydrophobic aliphatic chains into the cell membrane's lipid bilayer.
Ordering information: Contains a red tracer dye (50 µl), 4X labeling buffer (12,5 ml), and 10X HBSS (25 ml).
Delivery: Kit contains CYTO-ID® red tracer dye, 50 µl, 4X labeling buffer, 12,5 ml and 10X HBSS, 25 ml.