DNA methylation kit, 5-hmC, BIOARRAY™
BIOARRAY™ 5-hmC methylation kit, 50 Tests
Codice catalogo ENZOENZ450110050
Nome del fornitore: Enzo Life Sciences
DNA methylation kit, 5-hmC, BIOARRAY™
Codice catalogo ENZOENZ450110050
Nome del fornitore: Enzo Life Sciences
Numero CAS:
Vai a
Product Details & Documents
BIOARRAY™ 5-hmC Methylation Kit is used for the sequence specific detection of 5-hydroxymethylcytosine within DNA using a simple and efficient glucosylation reaction procedure.
- Single tube reaction format
- Suitable for analysis by a variety of downstream applications
- Flexible set-up for custom assay design
Informazioni per l'ordine: Kit contains 5-hmC Glucosyltransferase (5-hmC), 10X 5-hmC reaction buffer, 10X UDPG (uridine diphosphoglucose), MspI restriction enzyme, 5-hmC control DNA, qPCR primer 1, qPCR primer 2 and 5-DNA clean up concentrator kit.
Specifiche
Specifiche
- Conf:50 Tests
- Descrizione:BIOARRAY™ 5-hmC methylation kit