Thermal transfer printer labels, type B-499
: Brady
Printable, nylon cloth labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL recognised with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
Printable, nylon cloth labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL recognised with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
Product Family Options
- DescrizioneTipoModelloMaterialeAdesivoPrinting methodColoreStampanteLarghezzaDimensioni
- Printer labelsB-499THT-151-499-1.5-SCNylonAcrilico permanenteThermal transferbiancoBBP1250,80 mm6×50 mm1 * 1 ROUSFr. 264.00
- Printer labelsB-499THT-143-499-3NylonAcrilico permanenteThermal transferbianco360X-Plus II9 mm1 * 1 ROUSFr. 299.00
- Printer labelsB-499BPT-602-499NylonAcrilico permanenteThermal transferbiancoBBP129 mm1 * 1 ROUSFr. 291.00
- Printer labelsB-499BPT-602-499YLNylonAcrilico permanenteThermal transfergialloBBP129 mm1 * 1 ROUSFr. 335.00
- Printer labelsB-499THT-152-499-1.5-SCNylonAcrilico permanenteThermal transferbiancoBBP1225,40 mm9×25 mm1 * 1 ROUSFr. 264.00
- Printer labelsB-499THT-144-499-3NylonAcrilico permanenteThermal transferbianco360X-Plus II12 mm1 * 1 ROUSFr. 451.00
- Printer labelsB-499BPT-603-499NylonAcrilico permanenteThermal transferbiancoBBP1212 mm1 * 1 ROUSFr. 319.00
- Printer labelsB-499THT-68-499-10NylonAcrilico permanenteThermal transferbianco360X-Plus II12×19 mm1 * 1 ROUSFr. 661.00
- Printer labelsB-499THT-125-499-1.5-SCNylonAcrilico permanenteThermal transferbiancoBBP1222,86 mm12×22 mm1 * 1.500 STSFr. 264.00
- Printer labelsB-499BPT-611-499NylonAcrilico permanenteThermal transferbiancoBBP1212×25 mm1 * 1 ROUSFr. 486.00
- Printer labelsB-499BPT-620-499NylonAcrilico permanenteThermal transferbiancoBBP1212×25 mm1 * 1 ROUSFr. 274.00
- Printer labelsB-499THT-144-499-1.5-SCNylonAcrilico permanenteThermal transferbiancoBBP1248,26 mm12×48 mm1 * 1.500 STSFr. 308.00
- Printer labelsB-499THT-136-499-1.5-SCNylonAcrilico permanenteThermal transferbiancoBBP1238,10 mm19×38 mm1 * 1 ROUSFr. 264.00
- Printer labelsB-499THT-69-499-3NylonAcrilico permanenteThermal transferbianco360X-Plus II20×36 mm1 * 1 ROUSFr. 480.00
- Printer labelsB-499THT-126-499-3NylonAcrilico permanenteThermal transferbianco360X-Plus II22×12 mm1 * 1 ROUSFr. 429.00
- Printer labelsB-499BPT-618-499NylonAcrilico permanenteThermal transferbiancoBBP1222×38 mm1 * 1 ROUSFr. 851.00
- Printer labelsB-499THT-163-499-3NylonAcrilico permanenteThermal transferbianco360X-Plus II25×9 mm1 * 1 ROUSFr. 429.00
- Printer labelsB-499THT-152-499-3NylonAcrilico permanenteThermal transferbianco360X-Plus II25×9 mm1 * 1 ROUSFr. 429.00
- Printer labelsB-499BPT-659-499NylonAcrilico permanenteThermal transferbiancoBBP1225×12 mm1 * 1 ROUSFr. 466.00
- Printer labelsB-499BPT-621-499NylonAcrilico permanenteThermal transferbiancoBBP1225×36 mm1 * 1 ROUSFr. 851.00
- Printer labelsB-499THT-137-499-1.5-SCNylonAcrilico permanenteThermal transferbiancoBBP1250,80 mm25×50 mm1 * 1.500 STSFr. 341.00
- Printer labelsB-499BPT-608-499NylonAcrilico permanenteThermal transferbiancoBBP1241×15 mm1 * 1 ROUSFr. 411.00
- Printer labelsB-499THT-151-499-3NylonAcrilico permanenteThermal transferbianco360X-Plus II50×6 mm1 * 1 ROUSFr. 429.00
- Printer labelsB-499BPT-603-499YLNylonAcrilico permanenteThermal transfergialloBBP1212 mm1 * 1 ROUSFr. 365.00