Thermal transfer printer labels, type B-490, FreezerBondz
Informazioni su questo articolo
FreezerBondz thermal transfer printers labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen. These labels are on a 1" core, a 'smaller core' than the majority of labels. Recommended printer for these is the BBP11-34L printer.
- Good print smudge resistance
- Solvent resistance
- Good high and low temperature performance
- Widely used in liquid nitrogen and autoclave applications
Freezerbondz™ printable labels for thermal transfer printers. Freezerbondz™ labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen. They have good print smudge resistance, solvent resistance, and good high and low temperature performance. Freezerbondz™ labels perform well in common laboratory environments such as liquid nitrogen and autoclave applications. For tube/vial applications the label must be wrapped upon itself with at least 3 mm overlap.
10 varianti
Product Details & Documents
FreezerBondz thermal transfer printers labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen. These labels are on a 1" core, a 'smaller core' than the majority of labels. Recommended printer for these is the BBP11-34L printer.
- Good print smudge resistance
- Solvent resistance
- Good high and low temperature performance
- Widely used in liquid nitrogen and autoclave applications
Freezerbondz™ printable labels for thermal transfer printers. Freezerbondz™ labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen. They have good print smudge resistance, solvent resistance, and good high and low temperature performance. Freezerbondz™ labels perform well in common laboratory environments such as liquid nitrogen and autoclave applications. For tube/vial applications the label must be wrapped upon itself with at least 3 mm overlap.
Product Family Options
Informazioni sul prodotto
- TipoModelloMaterialeAdesivoPrinting methodColoreStampanteDimensioniConfezioneDisponibilitàPrice
- BRDY011263Printer labels, FreezerbondsPrinter labels, FreezerbondsB-490THT-154-490-3PoliestereAcrilico permanenteThermal transferbianco360X-Plus II31×9 mm3000 labels/rollLog in per disponibilità1 * 1 ROUSFr. 282.00
- BRDY011264Printer labels, FreezerbondsPrinter labels, FreezerbondsB-490THT-155-490-3PoliestereAcrilico permanenteThermal transferbianco360X-Plus II41×15 mm3000 labels/rollLog in per disponibilità1 * 1 ROUSFr. 319.00
- BRDY011265Printer labels, FreezerbondsPrinter labels, FreezerbondsB-490THT-156-490-3PoliestereAcrilico permanenteThermal transferbianco360X-Plus II48×25 mm3000 labels/rollLog in per disponibilità1 * 1 ROUSFr. 566.00
- BRDY114202Printer labelsPrinter labelsB-490THT-156-490-1.5-SCPoliestereAcrilico permanenteThermal transferbiancoBBP1248×25 mm1500 labels/rollLog in per disponibilità1 * 1 ROUSFr. 295.00
- BRDY011266Printer labelsPrinter labelsB-490THT-157-490-3PoliestereAcrilico permanenteThermal transferbianco360X-Plus II55×38 mm3000 labels/rollLog in per disponibilità1 * 1 ROUSFr. 848.00
- BRDY114203Printer labelsPrinter labelsB-490THT-157-490-1.5-SCPoliestereAcrilico permanenteThermal transferbiancoBBP1255×38 mm1500 labels/rollLog in per disponibilità1 * 1 ROUSFr. 424.00
- BRDY011267Printer labelsPrinter labelsB-490THT-158-490-3PoliestereAcrilico permanenteThermal transferbianco360X-Plus II95×38 mm3000 labels/rollLog in per disponibilità1 * 1 ROUSFr. 998.00
- BRDY114200Label printer tapeLabel printer tapeB-490THT-154-490-1.5-SCPoliestereAcrilico permanenteThermal transferbiancoBBP1231 mm×0,2 m1 roll/boxLog in per disponibilità1 * 1 ROUSFr. 141.00
- BRDY114201Label printer tapeLabel printer tapeB-490THT-155-490-1.5-SCPoliestereAcrilico permanenteThermal transferbiancoBBP1241 mm×0,3 m1 roll/boxLog in per disponibilità1 * 1 ROUSFr. 160.00
- BRDY114204Label printer tapeLabel printer tapeB-490THT-158-490-1.5-SCPoliestereAcrilico permanenteThermal transferbiancoBBP1295 mm×0,9 m1 roll/boxLog in per disponibilità1 * 1 ROUSFr. 499.00