Wafer tweezers, perforated
With 4 teeth, perforated lower paddle, 39S-4, 128 mm
Codice catalogo OUTI39S-4
Nome del fornitore: OUTILS RUBIS
Vai a
Product Details & Documents
Stainless steel. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers.
- Perforated lower paddle allows liquids to drop off
- Smooth polished non glare surface on the inside of the paddel in order to prevent chipping and marring the wafer
- Satin finish on the outside of the paddle for glare prevention
Specifiche
- Tipo:39S-4
- Descrizione:With 4 teeth, perforated lower paddle
- Lunghezza:128 mm
- Seq:10