Fluorescent nucleotides, Red 594 dUTP and Red 598 dUTP
Codice catalogo ENZOENZ42855
Nome del fornitore: ENZO LIFE SCIENCES
New Product
Numero CAS:
Specifiche
- Conf:25 Nmo
- Applicazione:
- Descrizione:Red 598 dUTP (1 mM)
- Seq:03
- Environmentally Preferable:
Specifiche
Informazioni su questo articolo
Red dUTP's (Red 594 dUTP, Red 598 dUTP) can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Fluorescently labelled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridisation procedures on fixed cells and tissues by direct fluorescence detection. Red dUTP's can also be used for multicolour fluorescence labelling.