Labels for BBP3x, type B-565
Supplier: Brady
Jump to:
About this item
These metallised polyester tapes are used for secure identification. Ideal for product rating plates, product warranty labels and asset identification labels.
3 Options Available Below
Product Details & Documents
These metallised polyester tapes are used for secure identification. Ideal for product rating plates, product warranty labels and asset identification labels.
Documents
Product Family Options
Product Information
- DescrizioneTipoModelloMaterialeAdesivoPrinting methodColoreStampanteDimensioniConfezioneAvailabilityPrice
- Metalised label printer tapeB-565B30C-1125-565-SLPoliestereAcrilico permanenteThermal transferargentoBBP3129 mm×30,4 m1 roll/boxLog in for availability1 * 1 ROUSFr. 327.00
- Metalised label printer tapeB-565B30C-2250-565-SLPoliestereAcrilico permanenteThermal transferargentoBBP3157 mm×30,4 m1 roll/boxLog in for availability1 * 1 ROUSFr. 523.00
- Metalised label printer tapeB-565B30C-4000-565-SLPoliestereAcrilico permanenteThermal transferargentoBBP31101 mm×30,4 m1 roll/boxLog in for availability1 * 1 ROUSFr. 818.00