Informazioni su questo articolo
This antibody recognizes a protein of ~35 kDa, identified as CD74. It is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74 has been shown to be useful in differentiating atypical fibroxanthoma (-) from malignant fibrous histiocytoma ( ).
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: LN-2 CLIP/813
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifiche
- Catalog No:
- BNC471207-500
- BNC471207-100
- Simbolo dell'antigene:
- CD74
- CD74
- Tipo anticorpo:
- Primary
- Primary
- Clonalità:
- Monoclonale
- Monoclonale
- Coniugazione:
- CF® 647
- CF® 647
- Clone:
- Clone LN-2 CLIP/813
- Clone LN-2 CLIP/813
- Reattività:
- Human
- Human
- Ospite:
- Mouse
- Mouse
- ID gene:
- 436568
- 436568
- Isotipo:
- IgG
- IgG
- ImmunoChimica:
- Yes
- Yes
- ImmunoFluorescenza:
- Yes
- Yes
- Concentrazione:
- 0,1 mg/ml
- 0,1 mg/ml
- Tampone di conservazione:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide