Anti-NLRP12 Rabbit Polyclonal Antibody [clone: ]
Catalog # ADIPAG25B0016C050
Supplier: AdipoGen Life Sciences
Specifications
- Tipo di anticorpi:NACHT, LRR And PYD Domains-Containing protein 12
- Simbolo dell'antigene:NLRP12
- Clonalità:Policlonale
- Ospite:Rabbit
- ImmunoPrecipitazione:Yes
- Reattività:Human
- Western blot:Yes
- Forma:Liquid
- Sinomini antigene:PAN6|RNO2|RNO|NALP12|PYPAF7|FCAS2|CLR19.3
- Tampone di conservazione:In PBS containing 10% glycerol and 0,02% sodium azide.
- Temperatura di conservazione:Short term: +4 °C, Long Term: –20 °C
- Concentrazione:0,5 mg/ml
- Immunogeno:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taglia:100 μg
- Conf:50 µG
Specifications
About this item
Type:
Antigen: NLRP12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human