Anti-ANGPTL6 Rabbit Polyclonal Antibody [clone: ]
ADIPAG25A0026C100
- Simbolo dell'antigene:ANGPTL6
- Clonalità:Policlonale
- ELISA:Yes
- Ospite:Rabbit
- Reattività:Human
- Western blot:Yes
- Forma:Liquid
- Tampone di conservazione:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Temperatura di conservazione:Short term: +4 °C, Long Term: –20 °C
- Concentrazione:1 mg/ml
- Immunogeno:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taglia:100 μg
- Conf:100 µG
Type:
Antigen: ANGPTL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human