Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-IDO Rabbit Polyclonal Antibody [clone: ]
Anti-IDO Rabbit Polyclonal Antibody [clone: ]
  ADIPAG25A0025C100
Anti-IDO Rabbit Polyclonal Antibody [clone: ]
  ADIPAG25A0025C100

 

  • Tipo di anticorpi:
    Indoleamine 2,3-dioxygenase
  • Simbolo dell'antigene:
    IDO
  • Clonalità:
    Policlonale
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Reattività:
    Human
  • Western blot:
    Yes
  • Forma:
    Liquid
  • Tampone di conservazione:
    0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
  • Temperatura di conservazione:
    Short term: +4 °C, Long Term: –20 °C
  • Concentrazione:
    1 mg/ml
  • Immunogeno:
    Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
  • Taglia:
    100 μg
  • Conf:
    100 µG

 

 

Type:
Antigen: IDO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human