Anti-NLRP3 Mouse Monoclonal Antibody [clone: Cryo-2]
Codice catalogo ADIPAG20B0012BC100
Nome del fornitore: AdipoGen Life Sciences
Specifiche
- Tipo di anticorpi:NLR Family, pyrin Domain Containing 3
- Simbolo dell'antigene:NLRP3
- Clonalità:Monoclonale
- Clone:Cryo-2
- Ospite:Mouse
- Isotipo:IgG2b
- Reattività:Human,Mouse
- Western blot:Yes
- Forma:Liquid
- Sinomini antigene:MWS|AII|LRR and PYD domains-containing protein 3|FCU|AGTAVPRL|cryopyrin|NACHT|and PYD-containing protein 3|NALP3|cold autoinflammatory syndrome 1 protein|C1orf7|CLR1.1|AVP|leucine-rich repeat-|CIAS1|nucleotide-binding oligomerization domain|FCAS|PYRIN-containing APAF1-like protein 1|leucine rich repeat and pyrin domain containing 3|PYPAF1|caterpiller protein 1.1|LRR and PYD containing protein 3|NACHT domain-
- Tampone di conservazione:In PBS containing 10% glycerol and 0,02% sodium azide.
- Temperatura di conservazione:Short term: +4 °C, Long Term: –20 °C
- Concentrazione:1 mg/ml
- Immunogeno:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taglia:100 μg
- Conf:100 µG
Specifiche
Informazioni su questo articolo
Type:
Antigen: NLRP3
Clonality: Monoclonal
Clone: Cryo-2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse