Anti-FGF-21 Mouse Monoclonal Antibody [clone: FG204-3]
Nome del fornitore: AdipoGen Life Sciences
Certificati
Informazioni su questo articolo
Type:
Antigen: FGF-21
Clonality: Monoclonal
Clone: FG204-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifiche
- Catalog No:
- ADIPAG20A0066C100
- ADIPAG20A0066C050
- Simbolo dell'antigene:
- FGF-21
- FGF-21
- Clonalità:
- Monoclonale
- Monoclonale
- Clone:
- FG204-3
- FG204-3
- Reattività:
- Human
- Human
- Ospite:
- Mouse
- Mouse
- Isotipo:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogeno:
- Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Forma:
- Liquid
- Liquid
- Concentrazione:
- 1 mg/ml
- 1 mg/ml
- Tampone di conservazione:
- 0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- 0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Temperatura di conservazione:
- Short term: +4 °C, Long Term: –20 °C
- Short term: +4 °C, Long Term: –20 °C
Specifiche
Frequently Bought Together