Anti-ADIPOQ Mouse Monoclonal Antibody [clone: ADI 943]
Catalog # ADIPAG20A0053C100
Supplier: AdipoGen Life Sciences
Specifications
- Tipo di anticorpi:Adiponectin
- Simbolo dell'antigene:ADIPOQ
- Clonalità:Monoclonale
- Clone:ADI 943
- ELISA:Yes
- Ospite:Mouse
- Isotipo:IgG1 kappa
- Reattività:Human
- Forma:Liquid
- Sinomini antigene:APM1|APM-1|GBP28|Adipocyte Complement Related Protein of 30kDa|ADPN|ACDC|ACRP30|ADIPQTL1
- Tampone di conservazione:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Temperatura di conservazione:Short term: +4 °C, Long Term: –20 °C
- Concentrazione:1 mg/ml
- Immunogeno:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taglia:50 μg
- Conf:100 µG
Specifications
About this item
Type:
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: ADI 943
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human