Anti-GITR Rat Monoclonal Antibody [clone: MGIT 02]
ADIPAG20A0007C100
- Tipo di anticorpi:Tumor necrosis Factor Receptor superfamily member 18
- Simbolo dell'antigene:GITR
- Clonalità:Monoclonale
- Clone:MGIT 02
- ELISA:Yes
- Ospite:Rat
- Isotipo:IgG2a kappa
- Reattività:Mouse
- Forma:Liquid
- Sinomini antigene:CD357|Tumor necrosis factor receptor superfamily member 18|GITR|Glucocorticoid-induced TNFR-related protein|TNFRSF18|Activation-inducible TNFR family receptor|UNQ319|AITR|PRO364
- Tampone di conservazione:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Temperatura di conservazione:Short term: +4 °C, Long Term: –20 °C
- Concentrazione:1 mg/ml
- Immunogeno:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Taglia:100 μg
- Conf:100 µG
Type:
Antigen: GITR
Clonality: Monoclonal
Clone: MGIT 02
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse