Order Entry
Switzerland
ContactUsLinkComponent
Anti-BSG Mouse Monoclonal Antibody [clone: 5D8]
Anti-BSG Mouse Monoclonal Antibody [clone: 5D8]
Catalog # GNTXGTX84795
Supplier:  Genetex
Anti-BSG Mouse Monoclonal Antibody [clone: 5D8]
Catalog # GNTXGTX84795
Supplier:  Genetex

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    basigin (Ok blood group)
  • Simbolo dell'antigene:
    BSG
  • Clonalità:
    Monoclonale
  • Clone:
    5D8
  • Coniugazione:
    Non coniugato
  • Citometria a flusso:
    Yes
  • Spostamento del gel:
    Yes
  • Ospite:
    Mouse
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG2b
  • Reattività:
    Human
  • Western blot:
    Yes
  • ID gene:
    682
  • Sinomini antigene:
    CD147|TCSF|5F7|EMMPRIN|OK|M6
  • Tampone di conservazione:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Temperatura di conservazione:
    For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
  • Immunogeno:
    Full length human recombinant protein of human BSG(NP_001719) produced in HEK293T cell.
  • Purificazione:
    Purified from mouse ascites fluids by Protein A/G affinity chromatography
  • Taglia:
    100 μl
  • Conf:
    100 µl

Specifications

About this item

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 5D8
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human