Anti-GBP2 Mouse Monoclonal Antibody [clone: 1E5]
GNTXGTX84445
: Genetex
- Tipo anticorpo:Primary
- Tipo di anticorpi:Guanylate binding protein 2, interferon-inducible
- Simbolo dell'antigene:GBP2
- Clonalità:Monoclonale
- Clone:1E5
- Coniugazione:Non coniugato
- Citometria a flusso:Yes
- Spostamento del gel:Yes
- Ospite:Mouse
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- Isotipo:IgG2b
- Reattività:Human,Dog
- Western blot:Yes
- ID gene:2634
- Tampone di conservazione:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Temperatura di conservazione:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogeno:Protein expressed in 293T cell transfected with human GBP2 expression vector
- Purificazione:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Taglia:100 μl
- Conf:100 µl
Type: Primary
Antigen: GBP2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1E5
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Dog