Anti-PDCD1 Mouse Monoclonal Antibody [clone: MIH4]
GNTXGTX42144
: Genetex
- Tipo anticorpo:Primary
- Tipo di anticorpi:Programmed cell death 1
- Simbolo dell'antigene:PDCD1
- Clonalità:Monoclonale
- Clone:MIH4
- Coniugazione:Non coniugato
- Citometria a flusso:Yes
- Ospite:Mouse
- ImmunoChimica:Yes
- Isotipo:IgG1
- Reattività:Human
- ID gene:5133
- Sinomini antigene:PD-1|hPD-1|Programmed cell death 1|hPD-l|CD279|hSLE1|PD1|SLEB2
- Tampone di conservazione:Phosphate buffered saline containing 0,09% Sodium Azide (NaN3)
- Temperatura di conservazione:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purificazione:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Taglia:100 μg
- Conf:100 µG
Type: Primary
Antigen: PDCD1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: MIH4
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human