Order Entry
Switzerland
ContactUsLinkComponent
Anti-CALD1 Mouse Monoclonal Antibody [clone: 12B5]
Anti-CALD1 Mouse Monoclonal Antibody [clone: 12B5]
Catalog # GNTXGTX28247
Supplier:  Genetex
Anti-CALD1 Mouse Monoclonal Antibody [clone: 12B5]
Catalog # GNTXGTX28247
Supplier:  Genetex

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Caldesmon 1
  • Simbolo dell'antigene:
    CALD1
  • Clonalità:
    Monoclonale
  • Clone:
    12B5
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Mouse
  • Isotipo:
    IgG1
  • Reattività:
    Human,
    Duck
  • Western blot:
    Yes
  • ID gene:
    800
  • Sinomini antigene:
    HCAD|caldesmon 1|NAG22|L-CAD|CDM|H-CAD|LCAD
  • Tampone di conservazione:
    PBS with 0,1% sodium azide
  • Temperatura di conservazione:
    Keep as concentrated solution, aliquot and store at 4 °C. Do not freeze. We recommend to parafilm vials during storage to prevent evaporation of buffer.
  • Concentrazione:
    1,5 mg/ml
  • Immunogeno:
    Duck gizzard caldesmon (heavy chain).
  • Purificazione:
    Protein A purified
  • Taglia:
    100 μg
  • Conf:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 12B5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Duck