Order Entry
Switzerland
ContactUsLinkComponent
Anti-NF1 Mouse Monoclonal Antibody [clone: McNFn27a]
Anti-NF1 Mouse Monoclonal Antibody [clone: McNFn27a]
Catalog # GNTXGTX28131
Supplier:  Genetex
Anti-NF1 Mouse Monoclonal Antibody [clone: McNFn27a]
Catalog # GNTXGTX28131
Supplier:  Genetex

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Neurofibromin 1
  • Simbolo dell'antigene:
    NF1
  • Clonalità:
    Monoclonale
  • Clone:
    McNFn27a
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Mouse
  • ImmunoChimica:
    Yes
  • ImmunoPrecipitazione:
    Yes
  • Isotipo:
    IgG1
  • Reattività:
    Human,
    Rat,
    Mouse
  • ID gene:
    4763
  • Sinomini antigene:
    Neurofibromin|NF1|VRNF|NFNS|WSS|neurofibromin|DKFZp686J1293
  • Tampone di conservazione:
    Ascitic fluid with 0,1% sodium azide
  • Temperatura di conservazione:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentrazione:
    10 mg/ml
  • Immunogeno:
    Synthetic N-terminal peptide, corresponding to amino acids 27-41 of Human Neurofibromin.
  • Purificazione:
    Ascites
  • Taglia:
    50 μl
  • Conf:
    50 µl

Specifications

About this item

Type: Primary
Antigen: NF1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: McNFn27a
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat