Order Entry
Switzerland
ContactUsLinkComponent
Anti-IFNG Mouse Monoclonal Antibody [clone: CH8920]
Anti-IFNG Mouse Monoclonal Antibody [clone: CH8920]
Catalog # GNTXGTX18639
Supplier:  Genetex
Anti-IFNG Mouse Monoclonal Antibody [clone: CH8920]
Catalog # GNTXGTX18639
Supplier:  Genetex

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Interferon gamma
  • Simbolo dell'antigene:
    IFNG
  • Clonalità:
    Monoclonale
  • Clone:
    CH8920
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Mouse
  • Isotipo:
    IgG1
  • Reattività:
    Human
  • ID gene:
    3458
  • Sinomini antigene:
    IFG|IFI
  • Tampone di conservazione:
    0,01 Phosphate-buffered saline, pH 7.0
  • Temperatura di conservazione:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentrazione:
    1 mg/ml
  • Immunogeno:
    Recombinant full length protein.
  • Purificazione:
    Protein G purified
  • Taglia:
    250μg
  • Conf:
    250 µG

Specifications

About this item

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: CH8920
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human