Anti-PTPRE Rabbit Polyclonal Antibody
100 μl
Codice catalogo GNTXGTX102466
Marketsource
Nome del fornitore: Genetex
Anti-PTPRE Rabbit Polyclonal Antibody
Codice catalogo GNTXGTX102466
Nome del fornitore: Genetex
Marketsource
Vai a
Product Details & Documents
Type: Primary
Antigen: PTPRE
Clonality: Polyclonal
Clone: Unconjugated
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifiche
- Taglia:100 μl
- Reattività:Human
- Immunogeno:Recombinant protein encompassing a sequence within the center region of human TSG-6. The exact sequence is proprietary.
- Purificazione:Purified by antigen-affinity chromatography.
- Tampone di conservazione:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
- Coniugazione:Non coniugato
- Isotipo:IgG
- Cat. No.:GNTXGTX102466
- Sinomini antigene:PTPE|HPTPE|R-PTP-EPSILON
- Simbolo dell'antigene:PTPRE
- Clonalità:Policlonale
- Ospite:Rabbit
- ID gene:7130
- Tipo anticorpo:Primary
- Temperatura di conservazione:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Tipo di anticorpi:Protein tyrosine phosphatase receptor type E
- Concentrazione:0,16mg/ml
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



