Orders LinkContactUsLinkComponent
Cuvette monouso
Rectangular cell, 1,5 ml, 10 mm, PS
  VARI6610018700
 :   VARIAN
Cuvette monouso
  VARI6610018700
 :  VARIAN
Cuvette monouso
Cuvette monouso
Rectangular cell, 1,5 ml, 10 mm, PS
1 * 1 ST
Increase flexibility and capabilities of spectrophotometers thanks to certified calibration and installation cells for optimum optical performance in any application.
  • Cammino ottico:
    10 mm
  • Volume:
    1,5 ml
  • Materiale finestra:
    PS
  • Descrizione:
    Rectangular cell
Accessories
  •  
    Descrizione
  • VARI110648190
    Cell base
    1 * 1 ST
  •  
    Descrizione
  • VARI10048100
    Cell holder base
    1 * 1 ST
  •  
    Descrizione
  • VARI110260190
    Standard cell holder (10 mm) incl. cell lifter and cell height adjustment screw
    1 * 1 ST
  •  
    Descrizione
  • VARI110645090
    Porta microcelle
    1 * 1 ST
  •  
    Descrizione
  • VARI210125300
    Cell holder with variable path length (5, 10, 20, 30, 40, 50 mm)
    1 * 1 ST
  •  
    Descrizione
  • VARI6610018900
    Magnetic stirrer bars (7×2 mm)
    1 * 1 ST
  •  
    Descrizione
  • VARI7418000400
    Magnetic stirring bar, PTFE