Orders LinkContactUsLinkComponent
Quartz cuvettes
Quartz cuvettes
Certificates
Quartz cuvettes
15 Options Available Below
Product Details & Documents
Flow cuvettes with center height of 15 mm and screw fitting connection.
For 8453 UV/Visible spectrophotometers.
Product Family Options
Product Information
  • Descrizione
    Altezza
    Volume
    Cammino ottico
    Materiale finestra
    Availability
    Price
  • Flow cell with oval aperture
    39 mm
    40 µl
    1 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5063-6570
  • Flow cell with oval aperture
    40 mm
    40 µl
    1 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5065-9907
  • Flow cell with oval aperture
    39 mm
    80 µl
    2 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5063-6571
  • Flow cell with oval aperture
    39 mm
    200 µl
    5 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5063-6572
  • Flow cell with oval aperture
    39 mm
    430 µl
    10 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5063-6573
  • Flow cell with rectangular aperture
    35 mm
    6,2 µl
    0,1 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5188-8003
  • Flow cell with rectangular aperture
    35 mm
    12,4 µl
    0,2 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5188-8004
  • Flow cell with rectangular aperture
    35 mm
    31 µl
    0,5 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5188-8005
  • Flow cell with rectangular aperture
    35 mm
    62 µl
    1 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5061-3396
  • Flow cell with rectangular aperture
    35 mm
    124 µl
    2 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5061-3397
  • Flow cell with rectangular aperture
    35 mm
    230 µl
    5 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5065-9918
  • Flow cell with rectangular aperture
    35 mm
    160 µl
    10 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5062-2476
  • Flow cell with rectangular aperture
    35 mm
    390 µl
    10 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5061-3398
  • Flow cell with round aperture
    35 mm
    30 µl
    10 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL0100-1224
  • Flow cell with round aperture
    35 mm
    80 µl
    10 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL0100-1225
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HEWL5061-3389
    Spacer for 2 mm cell
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HEWL9301-1161
    Stirring bars, LxØ: 7x5 mm
    Log in for availability
    1 * 2 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HEWL5061-3388
    Spacer for 1 mm cell
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HEWL9300-0761
    Lens cleaning papers
    Log in for availability
    1 * 50 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HEWL5063-6577
    Vassoio per 16 celle spettrofotometriche
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • HEWL5061-3390
    Spacer for 5 mm cell