Safety air outlet filter
Catalog # JOURJR-S-20004
Supplier: VICI JOUR RESEARCH
Specifications
- Descrizione:Safety air outlet filter
- Materiale:PP
- Seq:06
- Connettore:1/4-28 valve to cap and female Luer filter connections
- Conf:1
Specifications
About this item
This safety air outlet filter regulates the pressure during helium sparging.