Concrete moisture tester, Tramex®
Codice catalogo SIMELI9011
Nome del fornitore: Industrial Physics
Specifiche
- Modello:Tramex
- Descrizione:Concrete moisture encounter
- Peso:298 g
- L×P×A:85×150×38 mm
- Conf:1
Specifiche
Informazioni su questo articolo
Tramex is a non-destructive instrument for measuring moisture levels in concrete and sand-cement floors.
- Measurements up to a few centimeters deep
- Can be taken without damaging the surface
For use when laying parquet floors, floor coverings or floor coatings.