Detectable spatulas, SteriPlast®
263 mm, 150 mm, blu
Codice catalogo 231-0574
Nome del fornitore: BURKLE
Vai a
Product Details & Documents
These detectable SteriPlast® spatulas, made of blue PS, are produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. Ideal for sampling powders, granulates and pastes.
- Can be detected by colour scanners, metal detectors or X-rays
- Individually packaged and sterilised by gamma rays
- Production, assembly and packaging according to cleanroom class 7
- Complies with EU food requirements and FDA regulations
These spatulas are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
Documents
Specifiche
- Colore:blu
- Lunghezza:263 mm
- Lunghezza lama:150 mm