Reference photographs for painted steel surfaces, SSPC-VIS 2
Codice catalogo SIMELD3056
Nome del fornitore: Industrial Physics
Specifiche
- Descrizione:Method for evaluating degree of rusting on painted steel surfaces, SSPC-VIS 2
- Conf:1
Specifiche
Informazioni su questo articolo
SSPC-VIS 2 pictorial provides a method of evaluating degree of rusting on painted steel surfaces.
- Series of full colour images
- 27 black and white diagrams
- Various degrees of spot, general and pinpoint rusting on painted steel surfaces
Text and tables provide a guide to the use of the reference photographs, scales and description of standard rust grades, comparison of SSPC, ASTM, ISO and other rust grade scales and other useful information.