Order Entry
Switzerland
ContactUsLinkComponent
Anti-SELP Mouse Monoclonal Antibody (FITC) [clone: P.seK02.22]
Anti-SELP Mouse Monoclonal Antibody (FITC) [clone: P.seK02.22]
Catalog # ABNOMAB13966
Supplier:  Abnova
Anti-SELP Mouse Monoclonal Antibody (FITC) [clone: P.seK02.22]
Catalog # ABNOMAB13966
Supplier:  Abnova

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Selectin P
  • Simbolo dell'antigene:
    SELP
  • Clonalità:
    Monoclonale
  • Clone:
    P.seK02.22
  • Coniugazione:
    FITC (Fluoresceina Isotiocionato)
  • Ospite:
    Mouse
  • Isotipo:
    IgG2b
  • Reattività:
    Human
  • ID gene:
    6403
  • Taglia:
    100 reactions
  • Conf:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against human SELP.

Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: P.seK02.22
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human