Cleanroom mops, standard, MicroMopp CR/A
Catalog # PPSA3500067
Supplier: PFENNIG REINIGUNGSTECHNIK
Specifications
- Descrizione:MicroMopp Standard CR/A mops, 110 g
- Lunghezza:400 mm
- Conf:1
Specifications
About this item
MicroMopp Standard CR/A mops are special multiple-use cleanroom mops.
- Completely closed processing of polyester microsnares
- 100 % PES-microfiber with polypropylene bristle
- Rectangular mop design to reach edges perfectly well
- Very good slide characteristics, not suitable for chlorinated disinfectants
- For dissolving adhering contaminations and suitable for impregnation with flat wringer
- With water-permeable cover and PU backfilling four enormous sqm-performance
- Polyester pocket with drainage function