Test kits, sodium hypochlorite, Comparator 2000+
Sodium hypochlorite test kit, 2 - 18% NaOCl, AF 139, 1 KIT
Codice catalogo LOVI411390
Nome del fornitore: Lovibond
Test kits, sodium hypochlorite, Comparator 2000+
Codice catalogo LOVI411390
Nome del fornitore: Lovibond
Numero CAS:
Vai a
Product Details & Documents
- Stable and accurate
- Robust, compact and portable
- Prism enabling colour matching
- Diffuser plate is used to measure the turbidity
- Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
Delivery information: Kit is supplied in a robust carrying case containing Comparator 2000+, sodium hypochlorite comparator disc, 2× comparator vials, 10 ml, 13 mm path length, tablet reagents, crushing / stirring rod, 2× shaker tubes, plastic syringes 1 and 5 ml, vial cleaning brush and instruction sheet.
Specifiche
Specifiche
- Conf:1 KIT
- Descrizione:Sodium hypochlorite test kit
- Tipo:AF 139
- Range di misura:2 - 18% NaOCl