Orders LinkContactUsLinkComponent
Sampling cannulae
Sampling cannulae
Sampling cannulae
Sampling cannulae
Sampling cannulae
Standard cannula are unique, proprietary configurations for most manual and automated sampling applications.
  • Manufactured in both electropolished 316 stainless steel or PEEK
  • Can accommodate all standard tip filters
  • Optional stoppers set the proper sampling position
The USP defines the sampling point as: 'A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used.'
Product Family Options
Agilent/VanKel/Varian compatible cannulae
  • Descrizione
    Materiale
    Lunghezza
    Ø×L
  • Bent cannula with Luer lock
    Acciaio inossidabile 316
    3,2×120 mm
    1 * 1 ST
    1 * 1 ST
    Specifications
    Cat. No.
    QLAACAN475-01
  • Bent cannula with Luer lock
    PEEK
    3,2×120 mm
    1 * 1 ST
    1 * 1 ST - SFr. 261.00
    SFr. 261.00
    1 * 1 ST
       SFr. 261.00
    Specifications
    Cat. No.
    QLAACA8475-PV
Sotax compatible cannulae
  • Descrizione
    Materiale
    Lunghezza
    Ø×L
  • Bent cannula with Luer lock
    Acciaio inossidabile 316
    330 mm
    1 * 1 ST
    1 * 1 ST - SFr. 438.00
    SFr. 438.00
    1 * 1 ST
       SFr. 438.00
    Specifications
    Cat. No.
    QLAACAN013-ST-9
Hanson compatible cannulae
  • Descrizione
    Materiale
    Lunghezza
    Ø×L
  • Bent cannula with Luer lock
    PEEK
    3,2×120 mm
    1 * 1 ST
    1 * 1 ST - SFr. 259.00
    SFr. 259.00
    1 * 1 ST
       SFr. 259.00
    Specifications
    Cat. No.
    QLAACAN475-PH
  • Bent cannula with Luer lock
    PEEK
    3,2×195 mm
    1 * 1 ST
    1 * 1 ST - SFr. 256.00
    SFr. 256.00
    1 * 1 ST
       SFr. 256.00
    Specifications
    Cat. No.
    QLAACAN775-PH
Accessories
  •  
    Descrizione
  • QLAACANSTP-HR
    Stopper for adjustable cannula, Hanson, Ø = 3,2 mm
    1 * 1 ST