Illuminated surface microscopes
Supplier: Industrial Physics
About this item
Portable microscopes for inspectors to examine surface cleanliness, marks, pinholes, fish eyes, delaminating, cracks, and other coating defects on the spot.
- Both scale and lens with a focusing dial
- Surface to assess always clearly visible
- The image you see will be inverted
Delivery information: Supplied in a black leather case.
4 Options Available Below
Product Details & Documents
Portable microscopes for inspectors to examine surface cleanliness, marks, pinholes, fish eyes, delaminating, cracks, and other coating defects on the spot.
- Both scale and lens with a focusing dial
- Surface to assess always clearly visible
- The image you see will be inverted
Delivery information: Supplied in a black leather case.
Documents
TQC Company Video
Videos
Specifications for Product Family
Specifications
- Catalog No:
- SIMELD6169
- SIMELD6170
- SIMELD6172
- SIMELD6174
- Modello:
- LD6169
- LD6170
- LD6172
- LD6174
- L×P×A:
- 45×45×45 mm
- 175×90×55 mm
- 175×90×55 mm
- 175×90×55 mm
- Peso:
- 100 g
- 370 g
- 370 g
- 370 g
Specifications
Product Family Options
Product Information
- DescrizioneIngrandimentoAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SIMELD6169ModelloLD6169L×P×A45×45×45 mmPeso100 g - Specifications:
More
Specifications
Cat. No.SIMELD6170ModelloLD6170L×P×A175×90×55 mmPeso370 g - Specifications:
More
Specifications
Cat. No.SIMELD6172ModelloLD6172L×P×A175×90×55 mmPeso370 g - Illuminated microscope with 0,01 mm graduation scale100×Log in for availability1 * 1 STSFr. 714.00Specifications:
More
Specifications
Cat. No.SIMELD6174ModelloLD6174L×P×A175×90×55 mmPeso370 g