HPLC columns, C18
Nome del fornitore: FORTIS TECHNOLOGIES
Vai a
Informazioni su questo articolo
Fortis C18 is a pure silica based stationalry phase with unique high and low pH performance.
- pH range 1 to 12
- Provide superior peak shapes
- Fully scalable - UHPLC to prep
42 varianti
Product Details & Documents
Fortis C18 is a pure silica based stationalry phase with unique high and low pH performance.
- pH range 1 to 12
- Provide superior peak shapes
- Fully scalable - UHPLC to prep
Product Family Options
Informazioni sul prodotto
- Dimensioni particelleØ int.LunghezzaDisponibilitàPrice
- FORTF18-020101Fortis C18 HPLC columns
- FORTF18-020201Fortis C18 HPLC columns
- FORTF18-020301Fortis C18 HPLC columns
- HISMF18-020501Fortis C18 HPLC columns
- FORTF18-020701Fortis C18 HPLC columns
- FORTF18-030201Fortis C18 HPLC columns
- FORTF18-030301Fortis C18 HPLC columns
- FORTF18-030501Fortis C18 HPLC columns
- FORTF18-050201Fortis C18 HPLC columns
- FORTF18-050301Fortis C18 HPLC columns
- FORTF18-020102Fortis C18 HPLC columns
- FORTF18-020202Fortis C18 HPLC columns
- FORTF18-020302Fortis C18 HPLC columns
- FORTF18-020502Fortis C18 HPLC columns
- FORTF18-030202Fortis C18 HPLC columns
- FORTF18-030302Fortis C18 HPLC columns
- FORTF18-050302Fortis C18 HPLC columns
- FORTF18-020103Fortis C18 HPLC columns
- FORTF18-020203Fortis C18 HPLC columns
- FORTF18-020303Fortis C18 HPLC columns
- FORTF18-020503Fortis C18 HPLC columns
- FORTF18-020703Fortis C18 HPLC columns
- FORTF18-030203Fortis C18 HPLC columns
- FORTF18-030303Fortis C18 HPLC columns
- FORTF18-030503Fortis C18 HPLC columns
- FORTF18-030703Fortis C18 HPLC columns
- FORTF18-050303Fortis C18 HPLC columns
- FORTF18-050503Fortis C18 HPLC columns
- FORTF18-050703Fortis C18 HPLC columns
- FORTF18-050903Fortis C18 HPLC columns
- FORTF18-020205Fortis C18 HPLC columns
- FORTF18-020305Fortis C18 HPLC columns
- FORTF18-020505Fortis C18 HPLC columns
- FORTF18-020705Fortis C18 HPLC columns
- FORTF18-030205Fortis C18 HPLC columns
- FORTF18-030305Fortis C18 HPLC columns
- FORTF18-030505Fortis C18 HPLC columns
- FORTF18-030705Fortis C18 HPLC columns
- FORTF18-050305Fortis C18 HPLC columns
- FORTF18-050505Fortis C18 HPLC columns
- FORTF18-050705Fortis C18 HPLC columns
- FORTF18-050905Fortis C18 HPLC columns
This product has been blocked by your organization. Please contact your purchasing department for more information