Anion exchange columns, Dionex™, IonPac™ AS10
Supplier: DIONEX
Certificates
About this item
This IonPac® AS10 is a high-capacity hydroxide-selective, anion-exchange column designed for isocratic and gradient separation of inorganic anions and organic acids.
This column provides excellent resolution of weakly retained low-molecular-weight aliphatic acids. The selectivity and high capacity of the AS10 permits the analysis of trace-level analytes in high-ionic-strength matrices. The AS10 resin strongly retains nitrate, which makes this column well suited for the analysis of trace anions in nitric acid and nitrate salts.
Frequently Bought Together