Anion exchange columns, Dionex™, IonPac™ AS18
Nome del fornitore: DIONEX
Certificati
Informazioni su questo articolo
This high-capacity, high-efficiency column provides excellent resolution of inorganic anions in diverse sample matrices. This is the hydroxide-selective column of choice for fastest compliance monitoring of inorganic anions in drinking water and wastewater samples in accordance with U.S. EPA Methods 300.0 (A) and 300.1.
- Isocratic separation of common inorganic anions in 9 minutes
- Fast separation of the common inorganic anions in 4 min
- Superior retention and quantification of fluoride, acetate, and formate
These columns are recommended when separating the common inorganic anions and low-molecular weight organic acids in diverse sample matrices including drinking water, wastewater, leachates, foods, beverages, pharmaceutical preparations, and scrubber solutions.
Specifiche
- Catalog No:
- DION060549
- DION060551
- DION060553
- DION060555
- Pressione max.:
- 4000 psi
- 4000 psi
- 4000 psi
- 4000 psi
- Range di pH:
- 0 - 14
- 0 - 14
- 0 - 14
- 0 - 14