Orders LinkContactUsLinkComponent
Packed cell volume tubes (PCV), PS
Packed cell volume tubes (PCV), PS
Nome del fornitore:   TPP
Packed cell volume tubes (PCV), PS
Nome del fornitore:  TPP
Certificates
Packed cell volume tubes (PCV), PS
Packed cell volume tubes (PCV), PS
2 varianti
Product Details & Documents
These PCV tubes enables a quick and reproducible measuring technique of the cell volume.
  • Fits in micro-centrifuges
  • With calibrated capillary and volume graduation
  • Monitor cell growth (cell mass)
If during a production process many parameters need to be optimized, many samples accumulate at the same time. Here a quick analysis with the PCV tube and the read-out process with "easy read" is the method of the choice.
A short centrifugation step (1 minute at 2500 xg) is sufficient to form a pellet in the calibrated capillary. There the pellet can be read with the "easy read" measuring device.
Certifications: ISO 9001:2008.
Informazioni per l'ordine: Supplied with polyethylene caps.
Product Family Options
Informazioni sul prodotto
  • Capacità
    Altezza
    Ø
    Disponibilità
    Price
  • TPPA87005
    Packed cell volume tube, with volume scale
    Packed cell volume tube, with volume scale
    1 ml
    43 mm
    10,5 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 50 ST
    1 * 50 ST - 121.00 SFr.
    121.00 SFr.
    1 * 50 ST
       121.00 SFr.
  • TPPA87007
    Packed cell volume tube, without volume scale, neutral
    Packed cell volume tube, without volume scale, neutral
    1 ml
    43 mm
    10,5 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 50 ST
    1 * 50 ST - 76.00 SFr.
    76.00 SFr.
    1 * 50 ST
       76.00 SFr.
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Range di misura
    Materiale
    Disponibilità
    Price
  • Easy read measuring device for read-out process with PCV tubes
    Easy read measuring device for read-out process with PCV tubes
    TPPA87010
    0 - 5 µl
    Alluminio
    Log in per disponibilità
    1 * 1 ST - SFr. 509.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Cap for PCV tubes, PE, Ø×H: 13,5×7
    Cap for PCV tubes, PE, Ø×H: 13,5×7
    TPPA87008-150
    Log in per disponibilità
    1 * 150 ST - SFr. 33.60