Orders LinkContactUsLinkComponent
Cell counter, 'easy read'
Cell counter, 'easy read'
Easy read measuring device for read-out process with PCV tubes, 0 - 5 µl, Alluminio, 320×33×38 mm
Codice catalogo TPPA87010
Nome del fornitore:   TPP
Cell counter, 'easy read'
Codice catalogo TPPA87010
Nome del fornitore:  TPP
Cell counter, 'easy read'
Cell counter, 'easy read'
Easy read measuring device for read-out process with PCV tubes, 0 - 5 µl, Alluminio, 320×33×38 mm
Price
SFr. 509.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
These 'easy read' measuring device are fast, easy to use, reproducible and accurate alternative to commonly used methods for determination of a cell number density. The read-out process of the cell mass in the PCV tube with this requires neither special training nor sophisticated expensive machines.
  • Accurate and reproducible cell pellet measuring
  • Solid re-usable and easy handling measuring device
  • Read-out device with magnifying glass and cross line
Optimal with the use of PCV tubes.
Specifiche
  • Range di misura:
    0 - 5 µl
  • Descrizione:
    Easy read measuring device for read-out process with PCV tubes
  • Materiale:
    Alluminio
  • L×P×A:
    320×33×38 mm