Orders LinkContactUsLinkComponent
Spore suspensions
Spore suspensions
Spore suspensions
Supplier:  CROSSTEX
Certificates
Spore suspensions
3 Options Available Below
Product Details & Documents
Documents
Product Family Options
Product Information
  • Descrizione
    Availability
    Price
  • Bacillus subtilis, cell line 6633: spore suspension (14 month shelf life), population 106/0,1 ml - steam sterilisation process
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - SFr. 1’130.00
    SFr. 1’130.00
    1 * 10 mL
       SFr. 1’130.00
  • Bacillus subtilis, cell line 5230: spore suspension (15 month shelf life), population 108/0,1 ml - steam sterilisation process
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - SFr. 3’900.00
    SFr. 3’900.00
    1 * 10 mL
       SFr. 3’900.00
  • Bacillus atrophaeus, cell line 9372: spore suspension (24 month shelf life), population 108/0,1 ml - steam sterilisation process
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - SFr. 3’100.00
    SFr. 3’100.00
    1 * 10 mL
       SFr. 3’100.00