Labware value pack, Nalgene®
Labware value pack
Codice catalogo 212-0440
Nome del fornitore: Thermo Fisher Scientific
Labware value pack, Nalgene®
Codice catalogo 212-0440
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Labware Value Pack is a useful assortment of popular labware items used in every lab.
Serves as new lab starter or refresher pack
- Sets the standard for quality and superior performance with the highest quality materials
- Shatterproof and it provides a safer, longer equipment life at lower replacement costs than glass
Note: Remove jar closure and autoclave separately. LDPE Unitary™ wash bottles are not autoclavable. All other items in Value Pack are autoclavable.
Certifications: ASTM E1272, CFR 21, ISO 6706, Part 177.1520.
Informazioni per l'ordine: Includes six straight-sided jars (500 ml/PPCO), four Unitary™ wash bottles (500 ml/LDPE), four graduated cylinders (2 to 100 ml, 500 ml, 1 L/PPCO), five Griffin low form beakers (50, 100, 250, 600, 1000 ml/PPCO), Unwire™ blue half-rack for 30 mm tubes.
Specifiche
- Descrizione:Labware value pack
- Cat. No.:212-0440
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)