Heating tapes, IT-S45
Informazioni su questo articolo
Isopad IT-S45 is a preterminated, flexible heating tape consisting of glassfibre insulated, flexible fabric.
- Maximum operating temperature: 450 °C
- Width×Thickness: 30×5 mm (±10%)
- Maximum power per meter 250 W/m
The protective overbraid of the coiled heating conductors meets the requirements of Protection Class I. This heating tape is typically used for heating up and compensating for heat losses on short pipes and small sized structures. The heating tape is not moisture protected and must only be used in dry areas.
6 varianti
Product Details & Documents
Isopad IT-S45 is a preterminated, flexible heating tape consisting of glassfibre insulated, flexible fabric.
- Maximum operating temperature: 450 °C
- Width×Thickness: 30×5 mm (±10%)
- Maximum power per meter 250 W/m
The protective overbraid of the coiled heating conductors meets the requirements of Protection Class I. This heating tape is typically used for heating up and compensating for heat losses on short pipes and small sized structures. The heating tape is not moisture protected and must only be used in dry areas.
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- ISOP386552-000Heating tape IT-S45, 0,5 m length, 100 W
- 454-3001Heating tape IT-S45, 1 m length, 250 W
- ISOP051330-000Heating tape IT-S45, 1,5 m length, 375 W
- 454-3002Heating tape IT-S45, 2 m length, 500 W
- 454-3003Heating tape IT-S45, 3 m length, 750 W
- ISOP932450-000Heating tape IT-S45, 5 m length, 1250 W