Immobilised reductant column
Codice catalogo PIER77701
Nome del fornitore: Thermo Fisher Scientific
Numero CAS:
Specifiche
- Conf:2 ml
- Descrizione:Immobilised reductant column
Specifiche
Informazioni su questo articolo
Pierce™ Immobilised reductant column contains beaded resin to which an thiol-based reducing agent has been immobilised to enable solid-phase reduction of peptide and protein disulphide bonds.
- Eliminates soluble components which interfere with the assay of free thiol groups
- No need to remove reducing agent after the reaction
- Reduced protein, free of contaminating thiols, can be easily assayed using the BCA method
- Solid phase; protocols for both peptides and proteins
- Excellent protein recovery
- Regenerable matrix useable up to four times