Kit analitici, sodio in cuvetta in soluzioni nutritive per fertilizzazione per Spectroquant®, Supelco®
Codice catalogo 1.00885.0001
Nome del fornitore: Merck
Numero CAS:
Specifiche
- Metodo chimico:Ferro (III) tiocianato
- Conf:25 Tests
- Descrizione:Sodio in cuvetta in soluzioni nutritive per fertilizzazione
- M-Clarity quality level:MQ100
- Per strumento:Prove, Nova 30/60, Move
- Range di misura:10 - 300 mg/l Na
Specifiche
Informazioni su questo articolo
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.