Pierce™ Peroxidase Suppressor
Codice catalogo PIER35000
Nome del fornitore: Thermo Fisher Scientific
Numero CAS:
Specifiche
- Conf:100 ml
- Descrizione:Peroxidase suppressor
Specifiche
Informazioni su questo articolo
This peroxidase suppressor is a stable and easy-to-use inhibitor of endogenous peroxidase activity from cells and tissues.
- Stable and ready-to-use
- Effectively inhibits peroxidase activity
- No interference with antigen-antibody interactions
Designed to reduce background signal in HRP-based immunohistochemistry applications.
Caution: Store at 4 °C upon receipt.