Immobilisation kit for peptides, SulfoLink™, Pierce™
Catalog # PIER44999
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Conf:1 KIT
- Descrizione:SulfoLink™ Immobilisation Kit for Peptides, 5×2 ml, immobilise 0,1 - 1 mg of sulfhydryl-peptide per column
Specifications
About this item
Immobilisation Kit for Peptides, SulfoLink™ is a kit containing all the reagents for preparing the peptide sample for efficient immobilisation and columns containing SulfoLink™ resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification.
- Specific to sulphhydryls (-SH) groups
- Prepare and couple protein samples in two hours
- Flexible coupling conditions, use pH 7,5 to 9,0 aqueous buffers, organic solvent or denaturant
- Streamlined protocols for sample preparation, immobilisation, and affinity purification
- Immobilise 1 to 2 mg of peptide per 2 ml column of SulfoLink™ Coupling Resin
When incubated with a solution of peptide that contains reduced cysteine residues, the iodoacetyl groups of SulfoLink™ Resin react specifically and efficiently with the exposed sulphhydryls (-SH) to form covalent and irreversible thioether bonds that permanently attach the peptide to the resin. The result is a custom-made affinity resin for purification of antibodies, antigens and other molecules of interest.
Delivery: Kit include SulfoLink™ columns, 2 ml (5 columns), SulfoLink™ coupling buffer (120 ml), wash solution (120 ml), PBS pack (makes 500 ml), Bond-Breaker TCEP solution, neutral pH (0,5 ml), L-Cysteine·HCl (100 mg), and column accessories.